missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybutyl Acetate 97.0+%, TCI America™

Catalog No. A16755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16755G 5 g
A167525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16755G Supplier TCI America Supplier No. A16755G
Only null left

Chemical Identifiers

CAS 1173168-18-7
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD06797047
InChI Key ZWUWDFWEMWMTHX-UHFFFAOYSA-N
Synonym Acetic Acid 2-Methoxybutyl Ester
PubChem CID 14993724
IUPAC Name 2-methoxybutyl acetate
SMILES CCC(COC(=O)C)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxybutyl Acetate