missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxybenzoyl Chloride 98.0+%, TCI America™
Supplier: TCI America M0108100G
Specifications
| 2-Methoxybenzoyl Chloride | |
| Yellow | |
| C8H7ClO2 | |
| 100 g | |
| o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi | |
| COC1=CC=CC=C1C(=O)Cl | |
| 170.592 | |
| 170.59 | |
| Liquid |
| 21615-34-9 | |
| 140°C | |
| MFCD00000664 | |
| 1729 | |
| RZNHSEZOLFEFGB-UHFFFAOYSA-N | |
| 2-methoxybenzoyl chloride | |
| 88969 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 21615-34-9 | |
| 170.592 | |
| RZNHSEZOLFEFGB-UHFFFAOYSA-N | |
| 88969 | |
| COC1=CC=CC=C1C(=O)Cl |
| C8H7ClO2 | |
| MFCD00000664 | |
| o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi | |
| 2-methoxybenzoyl chloride |
Safety and Handling
TSCA : Yes