missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoyl Chloride 98.0+%, TCI America™

Catalog No. M0108500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0108500G 500 g
M010825G 25 g
M0108100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M0108500G Supplier TCI America Supplier No. M0108500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21615-34-9
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000664
InChI Key RZNHSEZOLFEFGB-UHFFFAOYSA-N
Synonym o-anisoyl chloride, methoxybenzoyl chloride, benzoyl chloride, 2-methoxy, o-methoxybenzoyl chloride, 2-methoxybenzoic acid chloride, 2-methoxybenzoylchloride, o-anisoylchloride, 2-anisoyl chloride, o-anisic acid chloride, acmc-1cbfi
PubChem CID 88969
IUPAC Name 2-methoxybenzoyl chloride
SMILES COC1=CC=CC=C1C(=O)Cl

Specifications

Color Yellow
Boiling Point 140°C
Quantity 500 g
UN Number 1729
Formula Weight 170.59
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxybenzoyl Chloride