missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxybenzeneboronic acid, 97%
Description
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.
Solubility
Insoluble in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 102°C to 106°C |
| Quantity | 1 g |
| Beilstein | 3030981 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 151.96 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Methoxybenzeneboronic acid |
Safety and Handling
| missing translation for 'healthHazard1' | H315-H319-H335 |
| missing translation for 'healthHazard2' | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| missing translation for 'healthHazard3' | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only