missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-beta-estradiol 97.0+%, TCI America™
$61.61 - $163.64
Chemical Identifiers
| CAS | 362-07-2 |
|---|---|
| Molecular Formula | C19H26O3 |
| Molecular Weight (g/mol) | 302.414 |
| MDL Number | MFCD00010489 |
| InChI Key | CQOQDQWUFQDJMK-SSTWWWIQSA-N |
| Synonym | 2-methoxyestradiol, panzem, 2-methoxyestradiol-17beta, 2-hydroxyestradol 2-methyl ether, estradiol, 2-methoxy, 2-hydroxyestradiol 2-methyl ether, pulmolar, 2-methoxyestra-1,3,5 10-triene-3,17beta-diol, panzem ncd, 2-methoxy-17beta-estradiol |
| PubChem CID | 66414 |
| ChEBI | CHEBI:28955 |
| IUPAC Name | (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol |
| SMILES | CC12CCC3C(C1CCC2O)CCC4=CC(=C(C=C34)OC)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M253025MG
|
TCI America
M253025MG |
25 mg |
Each for $61.61
|
|
|||||
|
M2530100MG
|
TCI America
M2530100MG |
100 mg |
Each for $163.64
|
|
|||||
Chemical Identifiers
| 362-07-2 | |
| 302.414 | |
| CQOQDQWUFQDJMK-SSTWWWIQSA-N | |
| 66414 | |
| (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol |
| C19H26O3 | |
| MFCD00010489 | |
| 2-methoxyestradiol, panzem, 2-methoxyestradiol-17beta, 2-hydroxyestradol 2-methyl ether, estradiol, 2-methoxy, 2-hydroxyestradiol 2-methyl ether, pulmolar, 2-methoxyestra-1,3,5 10-triene-3,17beta-diol, panzem ncd, 2-methoxy-17beta-estradiol | |
| CHEBI:28955 | |
| CC12CCC3C(C1CCC2O)CCC4=CC(=C(C=C34)OC)O |
Specifications
| 362-07-2 | |
| White-Yellow | |
| MFCD00010489 | |
| 2-methoxyestradiol, panzem, 2-methoxyestradiol-17beta, 2-hydroxyestradol 2-methyl ether, estradiol, 2-methoxy, 2-hydroxyestradiol 2-methyl ether, pulmolar, 2-methoxyestra-1,3,5 10-triene-3,17beta-diol, panzem ncd, 2-methoxy-17beta-estradiol | |
| CC12CCC3C(C1CCC2O)CCC4=CC(=C(C=C34)OC)O | |
| 302.414 | |
| CHEBI:28955 | |
| ≥97.0% (GC) | |
| 2-Methoxy-beta-estradiol |
| 187°C | |
| C19H26O3 | |
| 25 mg | |
| CQOQDQWUFQDJMK-SSTWWWIQSA-N | |
| (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol | |
| 66414 | |
| 302.41 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : KG7537500
TSCA : No