missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. M28325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28325G 5 g
M283225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28325G Supplier TCI America Supplier No. M28325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175277-45-9
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.126
MDL Number MFCD00153203
InChI Key KNIGTEGBOBDGKP-UHFFFAOYSA-N
PubChem CID 2775312
IUPAC Name 2-methoxy-5-(trifluoromethyl)pyridine
SMILES COC1=NC=C(C=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 177.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)pyridine