missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylpyridine 98.0+%, TCI America™

Catalog No. M28981G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M28981G Supplier TCI America Supplier No. M28981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13472-56-5
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD08703669
InChI Key NFQGQMBFMIIIOR-UHFFFAOYSA-N
Synonym 6-Methoxy-3-picoline
PubChem CID 3014746
IUPAC Name 2-methoxy-5-methylpyridine
SMILES CC1=CN=C(C=C1)OC

Specifications

Color Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-methylpyridine