missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylpyridine 98.0+%, TCI America™

Catalog No. p-7133080
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21901G 1 g
M21905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21901G Supplier TCI America Supplier No. M21901G

Chemical Identifiers

CAS 100848-70-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD06200799
InChI Key HGRXBKDKSYDWLD-UHFFFAOYSA-N
Synonym 2-Methoxy-4-picoline
PubChem CID 14223472
IUPAC Name 2-methoxy-4-methylpyridine
SMILES CC1=CC(=NC=C1)OC

Specifications

Color Yellow
Boiling Point 50°C
Quantity 1 g
UN Number 1993
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4-methylpyridine