missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™

Catalog No. p-7132819
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17145G 5 g
M171425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17145G Supplier TCI America Supplier No. M17145G

Chemical Identifiers

CAS 1195-66-0
Molecular Formula C7H15BO3
Molecular Weight (g/mol) 158.00
MDL Number MFCD03093086
InChI Key JZZJAWSMSXCSIB-UHFFFAOYSA-N
Synonym methoxyboronic acid pinacol ester, methoxyboronic acid, pinacol ester, 2-methoxy-4,4,5,5-tetramethyl 1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-methoxy-4,4,5,5-tetramethyl, zlchem 167, methyl pinacolyl borate, acmc-209a3a, ksc498e3h, methoxy boronic acid pinacol ester, 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborole
PubChem CID 10975759
IUPAC Name 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COB1OC(C)(C)C(C)(C)O1

Specifications

Color Colorless
Boiling Point 120°C
Quantity 5 g
UN Number 1993
Formula Weight 158.00
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane