missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 97%

Catalog No. AAL1905614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1905614 25 g
AAL1905606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1905614 Supplier Thermo Scientific Chemicals Supplier No. L1905614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1195-66-0 | C7H15BO3 | 158.00 g/mol

It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis, optical, and electrochemical properties of novel copolymers on the basis of benzothiadiazole and electron-rich arene units.

Notes
Moisture sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.

Chemical Identifiers

CAS 1195-66-0
Molecular Formula C7H15BO3
Molecular Weight (g/mol) 158.00
MDL Number MFCD03093086
InChI Key JZZJAWSMSXCSIB-UHFFFAOYSA-N
Synonym methoxyboronic acid pinacol ester, methoxyboronic acid, pinacol ester, 2-methoxy-4,4,5,5-tetramethyl 1,3,2-dioxaborolane, 1,3,2-dioxaborolane, 2-methoxy-4,4,5,5-tetramethyl, zlchem 167, methyl pinacolyl borate, acmc-209a3a, ksc498e3h, methoxy boronic acid pinacol ester, 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborole
PubChem CID 10975759
IUPAC Name 2-methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES COB1OC(C)(C)C(C)(C)O1

Specifications

Density 0.966
Boiling Point 120°C (228 mmHg)
Flash Point 46°C (114°F)
Refractive Index 1.41
Quantity 25 g
UN Number UN1993
Beilstein 2827758
Sensitivity Moisture sensitive
Formula Weight 158.01
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only