missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-pyridinecarboxaldehyde, 96%, Thermo Scientific™

Catalog No. p-4229281
Change view
Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439780050 5g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439780050 Supplier Thermo Scientific Chemicals Supplier No. 439780050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71255-09-9
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key PIFFMIDNNWOQLK-UHFFFAOYSA-N
Synonym 2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
PubChem CID 12533391
IUPAC Name 2-methoxypyridine-3-carbaldehyde
SMILES COC1=C(C=CC=N1)C=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Boiling Point 200°C to 201°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5g
Formula Weight 137.14
Percent Purity 96%
Chemical Name or Material 2-Methoxy-3-pyridinecarboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only