missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-methylpyrazine 98.0+%, TCI America™

Catalog No. p-7132376
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08105G 5 g
M081025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08105G Supplier TCI America Supplier No. M08105G
Only null left

Chemical Identifiers

CAS 2847-30-5
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00006127
InChI Key VKJIAEQRKBQLLA-UHFFFAOYSA-N
Synonym pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a
PubChem CID 17898
IUPAC Name 2-methoxy-3-methylpyrazine
SMILES CC1=NC=CN=C1OC

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
UN Number 1993
Formula Weight 124.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-3-methylpyrazine