Learn More
2-Methoxy-3-methylpyrazine, 99%
CAS: 2847-30-5 | C6H8N2O | 124.143 g/mol
Supplier: Thermo Scientific Chemicals L0806714
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Methoxy-3-methylpyrazine | |
| 1.07 | |
| 55°C (131°F) | |
| 1.5065 | |
| 25 g | |
| 878809 | |
| VKJIAEQRKBQLLA-UHFFFAOYSA-N | |
| 2-methoxy-3-methylpyrazine | |
| 17898 | |
| 99% |
| 2847-30-5 | |
| 159°C | |
| C6H8N2O | |
| MFCD00006127 | |
| UN1993 | |
| pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a | |
| CC1=NC=CN=C1OC | |
| 124.143 | |
| 124.14 |
Chemical Identifiers
| 2847-30-5 | |
| 124.143 | |
| VKJIAEQRKBQLLA-UHFFFAOYSA-N | |
| 17898 | |
| CC1=NC=CN=C1OC |
| C6H8N2O | |
| MFCD00006127 | |
| pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a | |
| 2-methoxy-3-methylpyrazine |
Safety and Handling
GHS H Statement
H301-H226-H315-H319
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c
H226-H302
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 220-651-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only