missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde 98.0+%, TCI America™

Catalog No. M10405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10405G 5 g
M104025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10405G Supplier TCI America Supplier No. M10405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5392-12-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004004
InChI Key YIQGLTKAOHRZOL-UHFFFAOYSA-N
Synonym 2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
PubChem CID 79352
IUPAC Name 2-methoxynaphthalene-1-carbaldehyde
SMILES COC1=C(C2=CC=CC=C2C=C1)C=O

Specifications

Melting Point 84°C
Boiling Point 205°C
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-1-naphthaldehyde