missing translation for 'onlineSavingsMsg'
Learn More

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol

Catalog No. p-7046227
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1352322 100 g
AAL1352336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1352322 Supplier Thermo Scientific Chemicals Supplier No. L1352322
Only null left

CAS: 5039-78-1 | C9H18ClNO2 | 207.698 g/mol

2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 5039-78-1
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00060097
InChI Key RRHXZLALVWBDKH-UHFFFAOYSA-M
Synonym polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
PubChem CID 78738
IUPAC Name trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
SMILES CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point -25°C
Density 1.105
Boiling Point >100°C
Refractive Index 1.466
Quantity 100 g
Beilstein 8168395
Solubility Information It is soluble in water.
Formula Weight 207.7
Concentration or Composition (by Analyte or Components) 72% aq. soln.
Chemical Name or Material 2-(Methacryloyloxy)ethyltrimethylammonium chloride, stabilized with 150 to 200ppm 4-methoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-733-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only