missing translation for 'onlineSavingsMsg'
Learn More

2-Mesitylenesulfonyl chloride, 99%

Catalog No. p-156058
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125590250 25 g Plastic bottle
AC125591000 100 g Plastic bottle
AC125595000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC125590250 Supplier Thermo Scientific Chemicals Supplier No. 125590250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 773-64-8 | C9H11ClO2S | 218.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 773-64-8
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.7
MDL Number MFCD00007434
InChI Key PVJZBZSCGJAWNG-UHFFFAOYSA-N
Synonym 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride
PubChem CID 13046
IUPAC Name 2,4,6-trimethylbenzenesulfonyl chloride
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53°C to 57°C
Color Gray to White
Boiling Point 150°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3)3C6H2SO2Cl
Quantity 25 g
Beilstein 11, 136
Fieser 01,661; 05,430; 06,625; 08,318
Solubility Information Solubility in water: reacts. Other solubilities: soluble in ether,toluene and dichloromethane
Formula Weight 218.7
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mesitylenesulfonyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
EINECSNumber 212-257-8
RTECSNumber DB8930000
TSCA TSCA

RUO – Research Use Only