missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mesitylenesulfonyl Chloride 99.0+%, TCI America™
$90.48 - $90.48
Chemical Identifiers
| CAS | 773-64-8 |
|---|---|
| Molecular Formula | C9H11ClO2S |
| Molecular Weight (g/mol) | 218.695 |
| MDL Number | MFCD00007434 |
| InChI Key | PVJZBZSCGJAWNG-UHFFFAOYSA-N |
| Synonym | 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride |
| PubChem CID | 13046 |
| IUPAC Name | 2,4,6-trimethylbenzenesulfonyl chloride |
| SMILES | CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C |
Chemical Identifiers
| 773-64-8 | |
| 218.695 | |
| PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
| 13046 | |
| CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C |
| C9H11ClO2S | |
| MFCD00007434 | |
| 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
| 2,4,6-trimethylbenzenesulfonyl chloride |
Specifications
| 773-64-8 | |
| White | |
| C9H11ClO2S | |
| 25 g | |
| 2-mesitylenesulfonyl chloride, mesitylenesulfonyl chloride, 2,4,6-trimethylbenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4,6-trimethyl, mesitylsulfonyl chloride, 2-mesitylenesulfonylchloride, mesitylene-2-sulphonyl chloride, 2,4,6-trimethyl-benzenesulfonyl chloride, 2,4,6-trimethylphenylsulfonyl chloride, mesitylsulfonylchloride | |
| CC1=CC(=C(C(=C1)C)S(=O)(=O)Cl)C | |
| 218.695 | |
| 218.70 | |
| Crystalline Powder |
| 56°C | |
| 150°C | |
| MFCD00007434 | |
| 3261 | |
| PVJZBZSCGJAWNG-UHFFFAOYSA-N | |
| 2,4,6-trimethylbenzenesulfonyl chloride | |
| 13046 | |
| ≥99.0% (T) | |
| 2-Mesitylenesulfonyl Chloride |
Safety and Handling
RTECSNumber : DB8930000
TSCA : No
Recommended Storage : Refrigerator