missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptothiazole 98.0+%, TCI America™
$85.75 - $288.94
Chemical Identifiers
| CAS | 82358-09-6 |
|---|---|
| Molecular Formula | C3H3NS2 |
| Molecular Weight (g/mol) | 117.18 |
| MDL Number | MFCD00022449 |
| InChI Key | OCVLSHAVSIYKLI-UHFFFAOYSA-N |
| Synonym | 2-mercaptothiazole, 2 3h-thiazolethione, 2-thiazolethiol, 1,3-thiazole-2-thiol, thiazoline-2-thione, 2-mercapto-1,3-thiazole, 3h-thiazole-2-thione, thiazole-2-thiol, 1,3-thiazole-2 3h-thione, .delta.4-thiazoline-2-thione |
| PubChem CID | 1201387 |
| IUPAC Name | 2,3-dihydro-1,3-thiazole-2-thione |
| SMILES | S=C1NC=CS1 |
Chemical Identifiers
| 82358-09-6 | |
| 117.18 | |
| OCVLSHAVSIYKLI-UHFFFAOYSA-N | |
| 1201387 | |
| S=C1NC=CS1 |
| C3H3NS2 | |
| MFCD00022449 | |
| 2-mercaptothiazole, 2 3h-thiazolethione, 2-thiazolethiol, 1,3-thiazole-2-thiol, thiazoline-2-thione, 2-mercapto-1,3-thiazole, 3h-thiazole-2-thione, thiazole-2-thiol, 1,3-thiazole-2 3h-thione, .delta.4-thiazoline-2-thione | |
| 2,3-dihydro-1,3-thiazole-2-thione |
Specifications
| 82358-09-6 | |
| Yellow | |
| MFCD00022449 | |
| 2-mercaptothiazole, 2 3h-thiazolethione, 2-thiazolethiol, 1,3-thiazole-2-thiol, thiazoline-2-thione, 2-mercapto-1,3-thiazole, 3h-thiazole-2-thione, thiazole-2-thiol, 1,3-thiazole-2 3h-thione, .delta.4-thiazoline-2-thione | |
| S=C1NC=CS1 | |
| 117.18 | |
| 117.18 | |
| Crystalline Powder |
| 80°C | |
| C3H3NS2 | |
| 1 g | |
| OCVLSHAVSIYKLI-UHFFFAOYSA-N | |
| 2,3-dihydro-1,3-thiazole-2-thione | |
| 1201387 | |
| ≥98.0% (GC,T) | |
| 2-Mercaptothiazole |
Safety and Handling
TSCA : No