missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyrimidine, 98%

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol

$287.43 - $287.43

Chemical Identifiers

CAS 1450-85-7
Molecular Formula C4H4N2S
Molecular Weight (g/mol) 112.15
MDL Number MFCD00006073
InChI Key HBCQSNAFLVXVAY-UHFFFAOYSA-N
Synonym 2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
PubChem CID 1550489
ChEBI CHEBI:53576
IUPAC Name 1H-pyrimidine-2-thione
SMILES C1=CNC(=S)N=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156740500
SDS View Documents
Thermo Scientific Chemicals
156740500
50 g Glass bottle
Each for $287.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1450-85-7
112.15
HBCQSNAFLVXVAY-UHFFFAOYSA-N
1550489
1H-pyrimidine-2-thione
C4H4N2S
MFCD00006073
2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
CHEBI:53576
C1=CNC(=S)N=C1

Specifications

1450-85-7
100.0
Yellow
98%
C4H4N2S
50 g
HBCQSNAFLVXVAY-UHFFFAOYSA-N
1H-pyrimidine-2-thione
1550489
112.15
Powder
97.5
230°C
Authentic
Glass bottle
MFCD00006073
2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
C1=CNC(=S)N=C1
112.15
CHEBI:53576
98%
2-Mercaptopyrimidine, 98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-917-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only