missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyrimidine, 98%

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol

Supplier:  Thermo Scientific Chemicals A1338222

 View more versions of this product

Catalog No. AAA1338222


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Mercaptopyrimidine
∼230°C (decomposition)
MFCD00006073
107105
HBCQSNAFLVXVAY-UHFFFAOYSA-N
1H-pyrimidine-2-thione
1550489
112.15
1450-85-7
C4H4N2S
100 g
2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
C1=CNC(=S)N=C1
112.15
CHEBI:53576
98%

Chemical Identifiers

1450-85-7
112.15
HBCQSNAFLVXVAY-UHFFFAOYSA-N
1550489
1H-pyrimidine-2-thione
C4H4N2S
MFCD00006073
2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
CHEBI:53576
C1=CNC(=S)N=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 215-917-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only