missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptopyridine N-Oxide Zinc Salt 98.0+%, TCI America™
$184.54 - $1258.24
Chemical Identifiers
| CAS | 13463-41-7 |
|---|---|
| Molecular Formula | C5H5NOS |
| MDL Number | MFCD00067336 |
Chemical Identifiers
| 13463-41-7 | |
| C5H5NOS | |
| MFCD00067336 |
Specifications
| 13463-41-7 | |
| C5H5NOS | |
| 25 g | |
| Bis(2-pyridylthio) Zinc 1,1′C-Dioxide, 1-Hydroxypyridine-2-thione Zinc Salt, 2-Pyridinethiol 1-Oxide Zinc Salt, Zinc Pyrithione | |
| ≥98.0% (T) | |
| 2-Mercaptopyridine N-Oxide Zinc Salt |
| White-Yellow | |
| MFCD00067336 | |
| 2811 | |
| 317.69 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3725&(9)-1110
RTECSNumber : ZH0950000
TSCA : Yes