missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine N-Oxide Zinc Salt 98.0+%, TCI America™

Catalog No. M063325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M063325G 25 g
M0633100G 100 g
M0633500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M063325G Supplier TCI America Supplier No. M063325G
Only null left

Chemical Identifiers

CAS 13463-41-7
Molecular Formula C5H5NOS
MDL Number MFCD00067336

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Synonym Bis(2-pyridylthio) Zinc 1,1′C-Dioxide, 1-Hydroxypyridine-2-thione Zinc Salt, 2-Pyridinethiol 1-Oxide Zinc Salt, Zinc Pyrithione
Formula Weight 317.69
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptopyridine N-Oxide Zinc Salt
EINECSNumber (5)-3725&(9)-1110
RTECSNumber ZH0950000
TSCA Yes