missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopropionic acid, 97%

Catalog No. p-7045319
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1025709 10 g
AAL1025722 100 g
AAL1025736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1025709 Supplier Thermo Scientific Chemicals Supplier No. L1025709
Only null left

CAS: 79-42-5 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79-42-5
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00004862
InChI Key PMNLUUOXGOOLSP-UHFFFAOYNA-N
Synonym 2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
PubChem CID 62326
ChEBI CHEBI:47872
IUPAC Name 2-sulfanylpropanoic acid
SMILES CC(S)C(O)=O

Specifications

Melting Point 10°C to 14°C
Density 1.196
Boiling Point 217°C
Flash Point 107°C (225°F)
Odor Unpleasant
Refractive Index 1.482
Quantity 10 g
UN Number UN2936
Beilstein 506218
Merck Index 14,9340
Formula Weight 106.14
Percent Purity 97%
Chemical Name or Material 2-Mercaptopropionic acid
Show More Show Less
Hazard Category H227-H290-H302-H314-H335
Hazard Statement GHS H Statement
H301-H331-H314-H318-H227
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P234-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THIOLACTIC ACID
EINECSNumber 201-206-5
RTECSNumber UF5250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only