missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich 2-Mercaptoethanol

Catalog No. M625010ML
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M625010ML 10 mL
M6250100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M625010ML Supplier Sigma Aldrich Supplier No. M625010ML
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2-mercaptoethanol is a thiol compound, commonly used as a reducing agent in organic reactions.bme is suitable for reducing protein disulfide bonds prior to polyacrylamide gel electrophoresis and is usually included in a sample buffer for sds-page at a concentration of 5%
  • Cleaving intermolecular (between subunits) disulfide bonds allows the subunits of a protein to separate independently on sds-page
  • Cleaving intramolecular (within subunit) disulfide bonds allows the subunits to become completely denatured so that each peptide migrates according to its chain length with no influence due to secondary structure

Specifications

CAS 60-24-2
Density 1.114 g/mL (at 25°C (literature))
Boiling Point 157°C (lit.)
Molecular Formula C2H6OS
Refractive Index n20/D 1.500 (literature)
Linear Formula HSCH2CH2OH
MDL Number MFCD00004890
Quantity 10 mL
Synonym beta-Mercaptoethanol; 2-Hydroxyethylmercaptan; BME; Thioethylene glycol
Molecular Weight (g/mol) 78.13
Percent Purity ≥99.0%
Show More Show Less