missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethane sulfonic acid sodium salt, 96%

Catalog No. p-7042852
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
25 g
Catalog No. Quantity
AAJ6398909 10 g
AAJ6398914 25 g
2 options

Catalog No. AAJ6398909

Supplier: Thermo Scientific Chemicals J6398909

Only null left

CAS: 19767-45-4 | C2H5NaO3S2 | 164.169 g/mol

2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptoethane sulfonic acid sodium salt is used as an adjuvant in cancer chemotherapy involving cyclophosphamide and ifosfamide. It is used as an antioxidant and cytoprotective agent in a wide variety of applications from protection from toxicity of therapeutic agents, such as cyclophosphamide to prevention from brain injury damage.

Solubility
Soluble in water. Also soluble in methanol, DMSO (33 mg/ml) and ethanol (<1mg/mL).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19767-45-4
Molecular Formula C2H5NaO3S2
Molecular Weight (g/mol) 164.169
MDL Number MFCD00007535
InChI Key XOGTZOOQQBDUSI-UHFFFAOYSA-M
Synonym mesna, sodium 2-mercaptoethanesulfonate, mesnex, uromitexan, mitexan, 2-mercaptoethanesulfonic acid sodium salt, mistabron, mesnum, mistabronco, mucofluid
PubChem CID 23662354
IUPAC Name sodium;2-sulfanylethanesulfonate
SMILES C(CS(=O)(=O)[O-])S.[Na+]

Specifications

Color White
Linear Formula HSCH2CH2SO3Na
Quantity 10 g
Beilstein 3657828
Merck Index 14,5909
Solubility Information Soluble in water. Also soluble in methanol,DMSO (33mg/ml) and ethanol (<1mg/ml).
Formula Weight 164.18
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Mercaptoethane sulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-285-9
RTECSNumber KI7968000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only