missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzoxazole 98.0+%, TCI America™

Catalog No. B009625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B009625G 25 g
B0096500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B009625G Supplier TCI America Supplier No. B009625G

Chemical Identifiers

CAS 2382-96-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00005769
InChI Key FLFWJIBUZQARMD-UHFFFAOYSA-N
Synonym 2-mercaptobenzoxazole, benzo d oxazole-2-thiol, 2-benzoxazolethiol, 2 3h-benzoxazolethione, benzoxazole-2-thiol, 2-benzoxazolinethione, 1,3-benzoxazole-2-thiol, benzoxazolinethione, 2-mercapto-1,3-benzoxazole, benzooxazole-2-thiol
PubChem CID 712377
IUPAC Name 3H-1,3-benzoxazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)O2

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 25 g
Formula Weight 151.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptobenzoxazole
EINECSNumber (5)-3395
RTECSNumber DM4882500
TSCA Yes