missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzoxazole, 98+%

Catalog No. p-7082079
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1234614 25 g
AAA1234622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1234614 Supplier Thermo Scientific Chemicals Supplier No. A1234614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2382-96-9 | C7H5NOS | 151.183 g/mol

2-Mercaptobenzoxazole is used in the surface treatment of materials. It is utilized for protection of metals from corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzoxazole is used in the surface treatment of materials. It is utilized for protection of metals from corrosion.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2382-96-9
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00005769
InChI Key FLFWJIBUZQARMD-UHFFFAOYSA-N
Synonym 2-mercaptobenzoxazole, benzo d oxazole-2-thiol, 2-benzoxazolethiol, 2 3h-benzoxazolethione, benzoxazole-2-thiol, 2-benzoxazolinethione, 1,3-benzoxazole-2-thiol, benzoxazolinethione, 2-mercapto-1,3-benzoxazole, benzooxazole-2-thiol
PubChem CID 712377
IUPAC Name 3H-1,3-benzoxazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)O2

Specifications

Melting Point 189°C to 194°C
Quantity 25 g
Beilstein 119483
Solubility Information Slightly soluble in water.
Formula Weight 151.19
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptobenzoxazole
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
EINECSNumber 219-191-9
RTECSNumber DM4882500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only