missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzimidazole 98.0+%, TCI America™
Supplier: TCI America B005525G
Specifications
| 2-Mercaptobenzimidazole | |
| 304°C | |
| C7H6N2S | |
| 25 g | |
| 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb | |
| S=C1NC2=CC=CC=C2N1 | |
| 150.20 | |
| 150.20 | |
| Crystalline Powder |
| 583-39-1 | |
| White | |
| MFCD00466107 | |
| 2811 | |
| YHMYGUUIMTVXNW-UHFFFAOYSA-N | |
| 2,3-dihydro-1H-1,3-benzodiazole-2-thione | |
| 707035 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 583-39-1 | |
| 150.20 | |
| YHMYGUUIMTVXNW-UHFFFAOYSA-N | |
| 707035 | |
| S=C1NC2=CC=CC=C2N1 |
| C7H6N2S | |
| MFCD00466107 | |
| 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb | |
| 2,3-dihydro-1H-1,3-benzodiazole-2-thione |
Safety and Handling
EINECSNumber : (5)-0472&(5)-0474
RTECSNumber : DE1050000
TSCA : Yes