missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole 98.0+%, TCI America™

Catalog No. B0055500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0055500G 500 g
B005525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0055500G Supplier TCI America Supplier No. B0055500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 583-39-1
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD00466107
InChI Key YHMYGUUIMTVXNW-UHFFFAOYSA-N
Synonym 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
PubChem CID 707035
IUPAC Name 2,3-dihydro-1H-1,3-benzodiazole-2-thione
SMILES S=C1NC2=CC=CC=C2N1

Specifications

Melting Point 304°C
Color White
Quantity 500 g
UN Number 2811
Formula Weight 150.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptobenzimidazole
EINECSNumber (5)-0472&(5)-0474
RTECSNumber DE1050000
TSCA Yes