missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole, 97%

Catalog No. AAA1835022
Change view
Click to view available options
Quantity:
100 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1835022 100 g
AAA183500E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1835022 Supplier Thermo Scientific Chemicals Supplier No. A1835022

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-39-1 | C7H6N2S | 150.20 g/mol

2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptobenzimidazole is used as an antidegradant. It is utilized for protecting rubber from oxidation. It is also employed as an intermediate in the preparation of rabeprazole. Further, it finds application in anti-leprosy drugs in medicine. In addition to this, it is used as a copper plating brightener.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 583-39-1
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD00466107
InChI Key YHMYGUUIMTVXNW-UHFFFAOYSA-N
PubChem CID 707035
SMILES S=C1NC2=CC=CC=C2N1

Specifications

Melting Point 305°C to 310°C
Flash Point >250°C (487°F)
Quantity 100 g
UN Number UN2811
Beilstein 119867
Merck Index 14,1082
Synonym 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
Solubility Information Soluble in alcohol
IUPAC Name 1,3-dihydrobenzimidazole-2-thione
Formula Weight 150.2
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzimidazole
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECSNumber DE1050000
TSCA TSCA

RUO – Research Use Only