Learn More
2-Mercapto-5-n-propylpyrimidine, 98%
CAS: 52767-84-7 | C7H10N2S | 154.23 g/mol
Supplier: Thermo Scientific Chemicals H2723906
Description
By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBy using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 2-Mercapto-5-n-propylpyrimidine | |
| 207°C to 211°C | |
| MFCD07777095 | |
| 5-propylpyrimidine-2-thiol, 2-mercapto-5-n-propylpyrimidine, pubchem9576, 5-propylpyrimidine-2 1h-thione | |
| CCCC1=CNC(=S)N=C1 | |
| 154.23 | |
| 154.24 |
| 52767-84-7 | |
| C7H10N2S | |
| 5 g | |
| YZFGTZRDMRKBBU-UHFFFAOYSA-N | |
| 5-propyl-1H-pyrimidine-2-thione | |
| 12356156 | |
| 98% |
Chemical Identifiers
| 52767-84-7 | |
| 154.23 | |
| YZFGTZRDMRKBBU-UHFFFAOYSA-N | |
| 12356156 |
| C7H10N2S | |
| MFCD07777095 | |
| 5-propylpyrimidine-2-thiol, 2-mercapto-5-n-propylpyrimidine, pubchem9576, 5-propylpyrimidine-2 1h-thione | |
| CCCC1=CNC(=S)N=C1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only