missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-n-propylpyrimidine, 98%

Catalog No. AAH2723903
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2723903 1 g
AAH2723906 5 g
AAH2723914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2723903 Supplier Thermo Scientific Chemicals Supplier No. H2723903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52767-84-7 | C7H10N2S | 154.23 g/mol

By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
By using 2-mercapto-5-n-propylpyrimidine (MPP) as capping ligands, copper nanoclusters with different core sizes were prepared using a chemical reduction method. 2-Mercapto-5-n-propylpyrimidine finds use as means of synthesizing enantiomerically enriched compounds. Used as an odorant to help detect natural gas leaks. As intermediate in organic syntheses.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 52767-84-7
Molecular Formula C7H10N2S
Molecular Weight (g/mol) 154.23
MDL Number MFCD07777095
InChI Key YZFGTZRDMRKBBU-UHFFFAOYSA-N
Synonym 5-propylpyrimidine-2-thiol, 2-mercapto-5-n-propylpyrimidine, pubchem9576, 5-propylpyrimidine-2 1h-thione
PubChem CID 12356156
SMILES CCCC1=CNC(=S)N=C1

Specifications

Melting Point 207°C to 211°C
Quantity 1 g
IUPAC Name 5-propyl-1H-pyrimidine-2-thione
Formula Weight 154.24
Percent Purity 98%
Chemical Name or Material 2-Mercapto-5-n-propylpyrimidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only