missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methylbenzimidazole, TCI America™

Catalog No. M135725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M135725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M135725G Supplier TCI America Supplier No. M135725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27231-36-3
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00010617
InChI Key CWIYBOJLSWJGKV-UHFFFAOYSA-N
Synonym 2-mercapto-5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole-2 3h-thione, 5-methyl-1h-benzimidazole-2-thiol, 5-methyl-2-benzimidazolethiol, 5-methyl-2-mercaptobenzimidazole, 5-methyl-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione, 1,3-dihydro-5-methyl, 1,3-dihydro-5-methyl-2h-benzimidazole-2-thione, 5-methyl-1h-benzoimidazole-2-thiol, 5-methylbenzimidazole-2-thiol
PubChem CID 712373
IUPAC Name 5-methyl-2,3-dihydro-1H-1,3-benzodiazole-2-thione
SMILES CC1=CC=C2NC(=S)NC2=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 164.23
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-5-methylbenzimidazole
EINECSNumber (5)-0473
TSCA Yes