missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methylbenzimidazole, 98%

Catalog No. AAA1119222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1119222 100 g
AAA1119206 5 g
AAA1119214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1119222 Supplier Thermo Scientific Chemicals Supplier No. A1119222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27231-36-3 | C8H8N2S | 164.23 g/mol

2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 27231-36-3
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00010617
InChI Key CWIYBOJLSWJGKV-UHFFFAOYSA-N
Synonym 2-mercapto-5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole-2 3h-thione, 5-methyl-1h-benzimidazole-2-thiol, 5-methyl-2-benzimidazolethiol, 5-methyl-2-mercaptobenzimidazole, 5-methyl-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione, 1,3-dihydro-5-methyl, 1,3-dihydro-5-methyl-2h-benzimidazole-2-thione, 5-methyl-1h-benzoimidazole-2-thiol, 5-methylbenzimidazole-2-thiol
PubChem CID 712373
IUPAC Name 5-methyl-1,3-dihydrobenzimidazole-2-thione
SMILES CC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point ∼295°C (decomposition)
Quantity 100 g
Beilstein 5083
Solubility Information Insoluble in water.
Formula Weight 164.23
Percent Purity 98%
Chemical Name or Material 2-Mercapto-5-methylbenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 248-350-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only