missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Mercapto-4-methyl-5-thiazolyl)acetic Acid 98.0+%, TCI America™
Supplier: TCI America M248125G
Specifications
| (2-Mercapto-4-methyl-5-thiazolyl)acetic Acid | |
| 211°C | |
| C6H7NO2S2 | |
| 25 g | |
| KYBOCQHDFLVQIB-UHFFFAOYSA-N | |
| 2-(4-methyl-2-sulfanylidene-3H-1,3-thiazol-5-yl)acetic acid | |
| 3035180 | |
| ≥98.0% (GC,T) |
| 34272-64-5 | |
| White-Yellow | |
| MFCD01750430 | |
| 5-(Carboxymethyl)-2-mercapto-4-methylthiazole | |
| CC1=C(SC(=S)N1)CC(=O)O | |
| 189.247 | |
| 189.25 | |
| Crystalline Powder |
Chemical Identifiers
| 34272-64-5 | |
| 189.247 | |
| KYBOCQHDFLVQIB-UHFFFAOYSA-N | |
| 3035180 | |
| CC1=C(SC(=S)N1)CC(=O)O |
| C6H7NO2S2 | |
| MFCD01750430 | |
| 5-(Carboxymethyl)-2-mercapto-4-methylthiazole | |
| 2-(4-methyl-2-sulfanylidene-3H-1,3-thiazol-5-yl)acetic acid |
Safety and Handling
RTECSNumber : XJ1921000
TSCA : No