missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-D-gluconic acid hemicalcium salt monohydrate, 99%

Catalog No. p-7043255
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Catalog No. Quantity
AAJ6660903 1 g
AAJ6660906 5 g
AAJ6660909 10 g
3 options

Catalog No. AAJ6660903

Supplier: Thermo Scientific Chemicals J6660903

Only null left

CAS: 3470-37-9 | C12H20CaO14 | 428.356 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3470-37-9
Molecular Formula C12H20CaO14
Molecular Weight (g/mol) 428.356
MDL Number MFCD00149104
InChI Key QACANIDDQFGAJU-JQTJLMCZSA-N
Synonym calcium 2-keto-d-gluconate
PubChem CID 131852723
IUPAC Name calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
SMILES C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]

Specifications

Color White
Quantity 1 g
Beilstein 3808734
Optical Rotation −70° (c=2 in Water)
Formula Weight 231.22
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Keto-D-gluconic acid hemicalcium salt monohydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only