missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropylnaphthalene 95.0+%, TCI America™
$108.11 - $108.11
Chemical Identifiers
| CAS | 2027-17-0 |
|---|---|
| Molecular Formula | C13H14 |
| Molecular Weight (g/mol) | 170.255 |
| MDL Number | MFCD00014322 |
| InChI Key | TVYVQNHYIHAJTD-UHFFFAOYSA-N |
| Synonym | 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene |
| PubChem CID | 16238 |
| IUPAC Name | 2-propan-2-ylnaphthalene |
| SMILES | CC(C)C1=CC2=CC=CC=C2C=C1 |
Chemical Identifiers
| 2027-17-0 | |
| 170.255 | |
| TVYVQNHYIHAJTD-UHFFFAOYSA-N | |
| 16238 | |
| CC(C)C1=CC2=CC=CC=C2C=C1 |
| C13H14 | |
| MFCD00014322 | |
| 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene | |
| 2-propan-2-ylnaphthalene |
Specifications
| 2027-17-0 | |
| 268°C | |
| MFCD00014322 | |
| 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene | |
| CC(C)C1=CC2=CC=CC=C2C=C1 | |
| 170.255 | |
| 170.26 | |
| Liquid |
| Yellow | |
| C13H14 | |
| 25 mL | |
| TVYVQNHYIHAJTD-UHFFFAOYSA-N | |
| 2-propan-2-ylnaphthalene | |
| 16238 | |
| ≥95.0% (GC) | |
| 2-Isopropylnaphthalene |
Safety and Handling
EINECSNumber : (4)-0314&(4)-0961
RTECSNumber : QJ8000000
TSCA : Yes