missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylnaphthalene, 96%

Catalog No. AAA1515306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1515306 5 g
AAA1515314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1515306 Supplier Thermo Scientific Chemicals Supplier No. A1515306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2027-17-0 | C13H14 | 170.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2027-17-0
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00014322
InChI Key TVYVQNHYIHAJTD-UHFFFAOYSA-N
Synonym 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene
PubChem CID 16238
IUPAC Name 2-propan-2-ylnaphthalene
SMILES CC(C)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 14°C
Density 0.975
Boiling Point 268°C
Flash Point 122°C (252°F)
Refractive Index 1.587
Quantity 5 g
Formula Weight 170.26
Percent Purity 96%
Chemical Name or Material 2-Isopropylnaphthalene
EINECSNumber 217-976-0
RTECSNumber QJ8000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only