missing translation for 'onlineSavingsMsg'
Learn More

2-(Isopropylamino)ethanol 99.0+%, TCI America™

Catalog No. I0227500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0227500ML 500 mL
I022725ML 25 mL
I0227100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0227500ML Supplier TCI America Supplier No. I0227500ML

Chemical Identifiers

CAS 109-56-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00041755
InChI Key RILLZYSZSDGYGV-UHFFFAOYSA-N
Synonym 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine
PubChem CID 7994
IUPAC Name 2-(propan-2-ylamino)ethanol
SMILES CC(C)NCCO

Specifications

Color Yellow
Boiling Point 171°C
Quantity 500 mL
Formula Weight 103.17
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Isopropylamino)ethanol
EINECSNumber (2)-0295
RTECSNumber KL5080000
TSCA Yes