missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-N,2,3-trimethylbutyramide 98.0+%, TCI America™

Catalog No. p-7131564
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I07295G 5 g
I072925G 25 g
2 options

Catalog No. I07295G

Supplier: TCI America I07295G

Only null left

Chemical Identifiers

CAS 51115-67-4
Molecular Formula C10H21NO
Molecular Weight (g/mol) 171.28
MDL Number MFCD00130070
InChI Key RWAXQWRDVUOOGG-UHFFFAOYSA-N
Synonym 2-isopropyl-n,2,3-trimethylbutyramide, 2-isopropyl-n,2,3-trimethylbutanamide, trimethyl isopropyl butanamide, methyl diisopropyl propionamide, n,2,3-trimethyl-2-1-methylethyl butanamide, unii-6qop5a9489, n,2,3-trimethyl-2-isopropylbutanamide, n,2,3-trimethyl-2-isopropylbutamide, butanamide, n,2,3-trimethyl-2-1-methylethyl, n,2,3-trimethyl-2-propan-2-yl butanamide
PubChem CID 65300
IUPAC Name N,2,3-trimethyl-2-(propan-2-yl)butanamide
SMILES CNC(=O)C(C)(C(C)C)C(C)C

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 85°C
Quantity 5 g
Formula Weight 171.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropyl-N,2,3-trimethylbutyramide