missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropyl-4-(N-methylaminomethyl)thiazole Dihydrochloride 98.0+%, TCI America™
$113.71 - $344.86
Chemical Identifiers
| CAS | 1185167-55-8 |
|---|---|
| Molecular Formula | C8H16Cl2N2S |
| Molecular Weight (g/mol) | 243.19 |
| MDL Number | MFCD11841218 |
| InChI Key | GNPOLJYMEXUGCG-UHFFFAOYSA-N |
| PubChem CID | 45595264 |
| IUPAC Name | N-methyl-1-(2-propan-2-yl-1,3-thiazol-4-yl)methanamine;dihydrochloride |
| SMILES | CC(C)C1=NC(=CS1)CNC.Cl.Cl |
Chemical Identifiers
| 1185167-55-8 | |
| 243.19 | |
| GNPOLJYMEXUGCG-UHFFFAOYSA-N | |
| N-methyl-1-(2-propan-2-yl-1,3-thiazol-4-yl)methanamine;dihydrochloride |
| C8H16Cl2N2S | |
| MFCD11841218 | |
| 45595264 | |
| CC(C)C1=NC(=CS1)CNC.Cl.Cl |
Specifications
| 1185167-55-8 | |
| MFCD11841218 | |
| GNPOLJYMEXUGCG-UHFFFAOYSA-N | |
| N-methyl-1-(2-propan-2-yl-1,3-thiazol-4-yl)methanamine;dihydrochloride | |
| 45595264 | |
| ≥98.0% (HPLC,T) | |
| 2-Isopropyl-4-(N-methylaminomethyl)thiazole Dihydrochloride |
| C8H16Cl2N2S | |
| 1 g | |
| CC(C)C1=NC(=CS1)CNC.Cl.Cl | |
| 243.19 | |
| 243.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator