missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-3-methoxypyrazine 98.0+%, TCI America™

Catalog No. I05771G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05771G 1 g
I05775G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05771G Supplier TCI America Supplier No. I05771G
Only null left

Chemical Identifiers

CAS 25773-40-4
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.20
MDL Number MFCD00006135
InChI Key NTOPKICPEQUPPH-UHFFFAOYSA-N
PubChem CID 33166
IUPAC Name 2-methoxy-3-(propan-2-yl)pyrazine
SMILES COC1=NC=CN=C1C(C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 152.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropyl-3-methoxypyrazine