missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-2-oxazoline 97.0+%, TCI America™

Catalog No. I06425G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I06425G Supplier TCI America Supplier No. I06425G
Only null left

Chemical Identifiers

CAS 10431-99-9
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD08276382
InChI Key FVEZUCIZWRDMSJ-UHFFFAOYSA-N
PubChem CID 566545
IUPAC Name 2-(propan-2-yl)-4,5-dihydro-1,3-oxazole
SMILES CC(C)C1=NCCO1

Specifications

Color Yellow
Boiling Point 138°C
Quantity 5 g
UN Number 1993
Formula Weight 113.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isopropyl-2-oxazoline