missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxypyridine-5-boronic acid, 97%

CAS: 870521-30-5 | C8H12BNO3 | 181.00 g/mol

$80.15 - $979.74

Chemical Identifiers

CAS 870521-30-5
Molecular Formula C8H12BNO3
Molecular Weight (g/mol) 181.00
MDL Number MFCD07368870
InChI Key SGEOBUJTURUUJE-UHFFFAOYSA-N
Synonym 6-isopropoxypyridin-3-yl boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxy-3-pyridyl boronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isopropoxy-5-pyridineboronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, 6-isoproxypyridine-3-boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
PubChem CID 44754864
IUPAC Name (6-propan-2-yloxypyridin-3-yl)boronic acid
SMILES CC(C)OC1=NC=C(C=C1)B(O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH64461MD
View Documents
Thermo Scientific Chemicals
H64461MD
250 mg N/A
Only null left
 
AAH6446106
View Documents
Thermo Scientific Chemicals
H6446106
5 g N/A
Only null left
 
Description

Description

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

870521-30-5
181.00
SGEOBUJTURUUJE-UHFFFAOYSA-N
44754864
CC(C)OC1=NC=C(C=C1)B(O)O
C8H12BNO3
MFCD07368870
6-isopropoxypyridin-3-yl boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxy-3-pyridyl boronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isopropoxy-5-pyridineboronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, 6-isoproxypyridine-3-boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
(6-propan-2-yloxypyridin-3-yl)boronic acid

Specifications

870521-30-5
MFCD07368870
6-isopropoxypyridin-3-yl boronic acid, 2-isoproxypyridine-5-boronic acid, 6-isopropoxypyridine-3-boronic acid, 6-isopropoxypyridin-3-ylboronic acid, 6-isopropoxy-3-pyridyl boronic acid, 2-isopropoxypyridine-5-boronic acid, 2-isopropoxy-5-pyridineboronic acid, 6-propan-2-yloxy pyridin-3-yl boronic acid, 6-isoproxypyridine-3-boronic acid, boronic acid, 6-1-methylethoxy-3-pyridinyl
SGEOBUJTURUUJE-UHFFFAOYSA-N
(6-propan-2-yloxypyridin-3-yl)boronic acid
44754864
97%
2-Isopropoxypyridine-5-boronic acid
C8H12BNO3
250 mg
Slightly soluble in water.
CC(C)OC1=NC=C(C=C1)B(O)O
181.00
181
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only