missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxyethanol, 99%

Catalog No. AAL13241AU
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13241AU 1000 mL
AAL13241AE 100 mL
AAL132410F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13241AU Supplier Thermo Scientific Chemicals Supplier No. L13241AU

CAS: 109-59-1 | C5H12O2 | 104.15 g/mol

2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 109-59-1
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00002866
InChI Key HCGFUIQPSOCUHI-UHFFFAOYSA-N
Synonym 2-isopropoxyethanol, isopropyl oxitol, isopropyl cellosolve, dowanal eipat, ethanol, 2-isopropoxy, isopropyl glycol, ethylene glycol isopropyl ether, isopropoxyethanol, ethanol, 2-1-methylethoxy, ucar ac
PubChem CID 7996
IUPAC Name 2-propan-2-yloxyethanol
SMILES CC(C)OCCO

Specifications

Melting Point -60°C
Density 0.903
Boiling Point 143°C
Flash Point 45°C (113°F)
Refractive Index 1.41
Linear Formula (CH3)2CHO(CH2)2OH
Quantity 1000 mL
UN Number UN1992
Beilstein 1732184
Solubility Information It is soluble in water.
Formula Weight 104.15
Percent Purity 99%
Chemical Name or Material 2-Isopropoxyethanol
Show More Show Less
Hazard Category H226-H312+H332-H319
Hazard Statement GHS H Statement
H226-H312-H332-H319
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-685-6
RTECSNumber KL5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only