missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxy-4,4,6-trimethyl-1,3,2-dioxaborinane 97.0+%, TCI America™

Catalog No. p-7131621
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08335G 5 g
I083325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08335G Supplier TCI America Supplier No. I08335G

Chemical Identifiers

CAS 61676-61-7
Molecular Formula C9H19BO3
Molecular Weight (g/mol) 186.058
MDL Number MFCD15143641
InChI Key AHCZPHINTQYKPU-UHFFFAOYSA-N
PubChem CID 12493145
IUPAC Name 4,4,6-trimethyl-2-propan-2-yloxy-1,3,2-dioxaborinane
SMILES B1(OC(CC(O1)(C)C)C)OC(C)C

Specifications

Color Colorless
Boiling Point 59°C
Quantity 5 g
UN Number 1993
Formula Weight 186.06
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Isopropoxy-4,4,6-trimethyl-1,3,2-dioxaborinane