missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 94%

Catalog No. AAL1727814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1727814 25 g
AAL1727806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1727814 Supplier Thermo Scientific Chemicals Supplier No. L1727814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61676-62-8 | C9H19BO3 | 186.058 g/mol

Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for fluorenylborolane preparation, reagent used to borylate arenes. Reagent used to borylate arenes and for fluorenylborolane preparation.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 61676-62-8
Molecular Formula C9H19BO3
Molecular Weight (g/mol) 186.058
MDL Number MFCD00192241
InChI Key MRWWWZLJWNIEEJ-UHFFFAOYSA-N
Synonym 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, isopropoxyboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane, pinbop, isopropoxyboronic acid, pinacol ester, isopropyl pinacol borate, 2-iso-propoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2-isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxa-borolane, 2-isopropoxy-4,4,5,5-dimethyl-1,3,2-dioxaborolane
PubChem CID 5017741
IUPAC Name 4,4,5,5-tetramethyl-2-propan-2-yloxy-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)OC(C)C

Specifications

Density 0.912
Boiling Point 73°C (15 mmHg)
Flash Point 42°C (107°F)
Refractive Index 1.409
Quantity 25 g
UN Number UN1993
Beilstein 1906370
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 186.06
Percent Purity 98%
Chemical Name or Material 2-Isopropoxy-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only