missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate, 96%

Catalog No. AC427670050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427670050 5 g Glass bottle
AC427670010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427670050 Supplier Thermo Scientific Chemicals Supplier No. 427670050
Only null left

CAS: 129946-88-9 | C14H8F6O3S2 | 402.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 129946-88-9
Molecular Formula C14H8F6O3S2
Molecular Weight (g/mol) 402.33
MDL Number MFCD00236132
InChI Key QXXHXTRTGZBOGD-UHFFFAOYSA-M
Synonym 5-trifluoromethyl-5h-dibenzo b,d thiophen-5-ium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, 5-trifluoromethyl dibenzothiophenium trifluoromethanesulfonate, s-trifluoromethyl dibenzothiophenium triflate, s-trifluoromethyl dibenzothiophenium trifluoromethanesulphonate, trifluoromethanesulfonate; 5-trifluoromethyl dibenzothiophen-5-ium, pubchem16425, acmc-20a6nt, c13h8f3s.chf3o3s, s-trifluoromethyl dibenzothiophenium trifluoromet
PubChem CID 2777507
IUPAC Name trifluoromethanesulfonate;5-(trifluoromethyl)dibenzothiophen-5-ium
SMILES [O-]S(=O)(=O)C(F)(F)F.FC(F)(F)[S+]1C2=CC=CC=C2C2=CC=CC=C12

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 150°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 402.33
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)dibenzothiophenium trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only