missing translation for 'onlineSavingsMsg'
Learn More

2-Isocyanatobenzonitrile, 97%, Thermo Scientific™

$392.48 - $392.48

Chemical Identifiers

CAS 42066-86-4
Molecular Formula C8H4N2O
Molecular Weight (g/mol) 144.133
InChI Key FQLNVWAFXQUXGW-UHFFFAOYSA-N
PubChem CID 2735873
IUPAC Name 2-isocyanatobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)N=C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB13667DA
SDS View Documents
Thermo Scientific Chemicals
BTB13667DA
1 g
Each for $392.48
Only null left
 
Description

Description

Specifications

Chemical Identifiers

42066-86-4
144.133
2735873
C1=CC=C(C(=C1)C#N)N=C=O
C8H4N2O
FQLNVWAFXQUXGW-UHFFFAOYSA-N
2-isocyanatobenzonitrile

Specifications

42066-86-4
Amber glass bottle
CNC6H4NCO
2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b
C1=CC=C(C(=C1)C#N)N=C=O
144.133
144.13
2-Isocyanatobenzonitrile
95%
C8H4N2O
1 g
FQLNVWAFXQUXGW-UHFFFAOYSA-N
2-isocyanatobenzonitrile
2735873
95%
Videos
SDS
Documents

Documents