missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-3-methylpyrazine 98.0+%, TCI America™

Catalog No. I03375ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03375ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I03375ML Supplier TCI America Supplier No. I03375ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13925-06-9
Molecular Formula C9H14N2
Molecular Weight (g/mol) 150.225
MDL Number MFCD00059770
InChI Key ZHMIODDNZRIENW-UHFFFAOYSA-N
Synonym 2-Methyl-3-isobutylpyrazine
PubChem CID 26333
IUPAC Name 2-methyl-3-(2-methylpropyl)pyrazine
SMILES CC1=NC=CN=C1CC(C)C

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 mL
Formula Weight 150.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-3-methylpyrazine